Learn More
Cytiva (formerly Pall Lab) Acro™ 50 Vent Devices with HDC II Media
Disposable filters ideal for the sterile venting of small containers and as an in-line gas filter.
Brand: Cytiva (formerly Pall Lab) 4672
Additional Details : Weight : 1.31500kg
Description
- 0.2 micron rated and contain a hydrophobic PTFE membrane
- The large effective filter area provides high flow rates and filters can be autoclaved several times ensuring low costs
Specifications
6 μm | |
Non-sterile | |
18/Pk. |