missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACP6 Antibody, HRP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99166H
Additional Details : Weight : 0.00970kg
Description
ACP6 Polyclonal specifically detects ACP6 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ACP6 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
acid phosphatase 6, lysophosphatidicPACPL1, Acid phosphatase-like protein 1, ACPL1acid phosphatase like 1, EC 3.1.3.2, LPAPlysophosphatidic acid phosphatase type 6 | |
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of human ACP6. | |
0.1 mL | |
Lipid and Metabolism | |
51205 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
HRP | |
PBS | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |