missing translation for 'onlineSavingsMsg'
Få mere at vide

Acifluorfen (90%), TRC

Artikelnummer. 30336384
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30336384 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30336384 Leverandør Toronto Research Chemicals Leverandørnr. TRCA795248250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Acifluorfen
CAS 50594-66-6
Molekylær formel C14 H7 Cl F3 N O5
Mængde 250 mg
Synonym 5-[2-Chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoic acid, 2-Nitro-5-(2-chloro-4-trifluoromethylphenoxy)benzoic acid, 3-Carboxy-2'-chloro-4'-trifluoromethyl-4-nitrodiphenyl ether, Acifluorfen, Acifluorfen H, Acifluorfene, PPG 847
InChI formel InChI=1S/C14H7ClF3NO5/c15-10-5-7(14(16,17)18)1-4-12(10)24-8-2-3-11(19(22)23)9(6-8)13(20)21/h1-6H,(H,20,21)
SMIL OC(=O)c1cc(Oc2ccc(cc2Cl)C(F)(F)F)ccc1[N+](=O)[O-]
IUPAC navn 5-[2-chloro-4-(trifluoromethyl)phenoxy]-2-nitrobenzoic acid
Molekylvægt (g/mol) 361.66
Formel vægt 360.9965
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.