missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Acid violet 17 (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acid Violet 17 |
| CAS | 4129-84-4 |
| Molekylær formel | C41 H44 N3 O6 S2 . Na |
| Mængde | 25 mg |
| Synonym | Benzenemethanaminium, N-[4-[[4-(diethylamino)phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-, hydroxide, inner salt, sodium salt, Benzenemethanaminium, N-[4-[[4-(diethylamino)phenyl][4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-, inner salt, sodium salt (9CI), C.I. Acid Violet 17 (6CI,7CI), C.I. Acid Violet 17, sodium salt (8CI), 1942 Violet, ADC Acid Brilliant Milling Violet 4BS, Acid Brilliant Milling Violet 4BS, Acid Leather Violet 2B, Acid Leather Violet 3B, Acid Leather Violet B, Acid Violet, Acid Violet 17, Acid Violet 4B, Acid Violet 4BN, Acid Violet 4BNS, Acid Violet 4BO, Acid Violet 4BS, Acid Violet 5BS, Acid Violet ACS, Acid Violet SR, Acidal Brilliant Violet 2B, Acidal Brilliant Violet 6B, Airedale Violet 4BNS, Amacid Violet 3B, Anadurm Violet M 2B, Brilliant Acid Milling Violet 4BS, Brilliant Acid Violet 4B, Brilliant Milling Violet S 4B, Brilliant Violet 5B, Bucacid Wool Violet, Burmese Violet 4BO, C.I. 42650, C.I. Food Violet 1, Calcocid Violet 4BXN, Cetil Brilliant Violet S 4B, Colocid Violet 4BH, Colocid Violet 5BN, Coomassie Lake Violet R, Coomassie Violet R, Coomassie Violet R 200, Covalene Violet S 4B, Derma Violet 4BR, Dinacid Violet 4BN, Dycosacid Violet 4BNS, Erio Violet 6BN, Fast Acid Violet B, Fenazo Violet X 4B, Food violet 1, Formyl Violet, Formyl Violet S 4B, Formyl Violet S 4BA, Hidacid Wool Violet, Hispacid Violet X 4BF, Indacid Violet 4BS, Kemacid Violet 4BS, Kenanthrol Violet 2B, Leather Violet S 4B, Libacid Violet L 4BS, Merantine Violet S 4B, Metamine Fast Violet 4B, Milling Brilliant Violet 2BR, Milling Violet S 4B, Mitsui Acid Violet 5BN, NSC 5019, NSC 71963, Naphthalene Leather Violet R, Neutral Violet, Orient Water Violet 7, Pontacyl Violet C 4BN, Pontacyl Violet S 4B, Ravi Acid Violet 4BN, Serva Violet 17, Shimazaki Acid Violet 5BN, Simacid Acid Violet, Solar Violet 4BN, Solar Violet 5BN, Special Violet S 4B-F, Sumitomo Acid Violet 5BN, Tertracid Milling Violet 4B, Triacid Violet S 4B, Triacor Violet S 4B, Vicoacid Violet BN, Violet 2EMBL, Violet 5BN, Violet S 4B, Violet S 4R, Vondamol Violet S 4B, Water Violet 178839, Water Violet 310503 |
| InChI formel | InChI=1S/C41H45N3O6S2.Na/c1-5-42(6-2)36-21-15-33(16-22-36)41(34-17-23-37(24-18-34)43(7-3)29-31-11-9-13-39(27-31)51(45,46)47)35-19-25-38(26-20-35)44(8-4)30-32-12-10-14-40(28-32)52(48,49)50;/h9-28H,5-8,29-30H2,1-4H3,(H-,45,46,47,48,49,50);/q;+1/p-1 |
| SMIL | [Na+].CCN(CC)c1ccc(cc1)C(=C2C=CC(=[N+](CC)Cc3cccc(c3)S(=O)(=O)[O-])C=C2)c4ccc(cc4)N(CC)Cc5cccc(c5)S(=O)(=O)[O-] |
| IUPAC navn | sodium;3-[[4-[[4-(diethylamino)phenyl]-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate |
| Molekylvægt (g/mol) | 761.92 |
| Formel vægt | 761.2569 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?