missing translation for 'onlineSavingsMsg'
Få mere at vide
Få mere at vide
Acid Red 88 (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acid Red 88 (Technical Grade) |
| CAS | 1658-56-6 |
| Molekylær formel | C20H13N2NaO4S |
| Mængde | 25 g |
| Synonym | C.I. Acid Red 88, Monosodium Salt, Naphthalene Red J, 11391 Red, 2-Naphthol Red J, Acid Cardinal G, Acid Fast Red A, Acid Leather Red ROC, Acid Red A, Acid Red A (Chinese), Acid Red AV, Acid Red G, Acid Rose AV, Acid Scarlet G, Airedale Red A, Amacid Fast Red A, Ambicid Fast Red E, Anadurm Red A-ROC, Anthrosin BRX, Apollo Acid Rocceline, Atul Acid Fast Red A, Azo Acid Red GS, Basacid Red 340, Benzyl Red ROC, Benzyl Red S, Brasilan Red S, Bucacid Fast Red A, C.I. 15620, C.I. Acid Red 88, Calcocid Fast Red A, Cavalene Red A, Colacid Red AV, Colocid Fast Red A, Conacid Red MM, Daedo Acid Roccelline NS, Dai-ei Roccelline, Derma Fur Red R 150, Diacid Red A, Dinacid Fast Red A, Dyacid Red J, Dycosacid Red A, Eniacid Fast Red A, Eriosin Roccelline, Eriosin Roccelline SS, Ext D and C Red No. 8Fabracid Red S-A, Fast Acid Red G, Fast Red A, Fast Red A (acid dye), Fast Red AE, Fast Red AG, Fast Red ALS, Fast Red AV, Fast Red MA, Fast Red S, Fenazo Red M, Fur Red KB, Hidacid Fast Red A, Hispacid Fast Red A, Ichoacid Red A, Indacid Acid Red A, Japan Red 506, Japan Red No. 506, Kayaku Roccelline, Kemacid Fast Red A, Libacid Fast Red A, Lurazol Red E, Monacid Red A, Multacid Red A, Multicuer Red A, Naphthalene Red JS, Naphtocard Fast Red C, Neklacid Fast Red A, Neutral Red R, New Red WO, Nitto Roccelline, Peony, Plastoresin Red RC, Pontacyl Fast Red AS, Red J, Red No. 506, Red for Furs G, Rifa Acid Roccelline NS, Rifa Leather Red E, Ritacid Rosalin AV, Rocceline, Roccelline, Roccelline A, Roccelline B, Roccelline G, Roccelline K, Roccelline KG, Roccelline L, Roccelline NS, Roccelline NS 200, Roccelline S, Rocelline, Romexal Red AV, Sandal Acid Red G, Shikiso Roccelline, Sodium 4-[(2-hydroxy-1-naphthyl)azo]-1-naphthalene Sulfonate, Tertracid Red RO, Toyo Roccelline, Triacid Fast Red Av, Triacid Red AN, Vicoacid Fast Red A, Vondacid Red GN, Vopsider Red AAV, 4-[(2-Hydroxy-1-naphthalenyl)azo]-1-naphthalenesulfonic Acid Monosodium Salt |
| InChI formel | InChI=1S/C20H14N2O4S.Na/c23-18-11-9-13-5-1-2-6-14(13)20(18)22-21-17-10-12-19(27(24,25)26)16-8-4-3-7-15(16)17;/h1-12,23H,(H,24,25,26);/q;+1/p-1/b22-21+; |
| SMIL | [Na+].Oc1ccc2ccccc2c1N=Nc3ccc(c4ccccc34)S(=O)(=O)[O-] |
| IUPAC navn | sodium;4-[(E)-(2-hydroxynaphthalen-1-yl)diazenyl]naphthalene-1-sulfonate |
| Molekylvægt (g/mol) | 400.38 |
| Formel vægt | 400.0494 |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?