Få mere at vide
Acid Orange 8 (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acid Orange 8 (Technical Grade) |
| CAS | 5850-86-2 |
| Molekylær formel | C17H13N2NaO4S |
| Mængde | 10 g |
| Synonym | 4-[(2-Hydroxy-1-naphthalenyl)azo]-3-methylbenzenesulfonic Acid Monosodium Salt, C.I. Acid Orange 8, monosodium salt (8CI)Acid Brilliant Orange RAcid Orange 8Acid Orange ROAiredale Orange RBasacid Orange 288Brilliant Orange RBrilliant Orange ROC.I. 15575C.I. Acid Orange 8Calcocid Orange RRDiacid Orange RKayaku Acid Orange ROKeyacid Orange ROKiton Fast Orange 2RKiton Orange RNaphthazine Orange ROrange 2ROOrange ROrange ROOrange TPolycor Orange RRitacid Orange ROShikiso Acid Orange ROSolar Orange RTertracid Orange ROTriacid Orange ROVondacid Orange ROWool Orange RYodogawa Acid Orange R |
| InChI formel | InChI=1S/C17H14N2O4S.Na/c1-11-10-13(24(21,22)23)7-8-15(11)18-19-17-14-5-3-2-4-12(14)6-9-16(17)20;/h2-10,20H,1H3,(H,21,22,23);/q;+1/p-1 |
| SMIL | [Na+].Cc1cc(ccc1N=Nc2c(O)ccc3ccccc23)S(=O)(=O)[O-] |
| IUPAC navn | sodium;4-[(E)-(2-hydroxynaphthalen-1-yl)diazenyl]-3-methylbenzenesulfonate |
| Molekylvægt (g/mol) | 364.35 |
| Formel vægt | 364.0494 |