missing translation for 'onlineSavingsMsg'
Få mere at vide

Acid Orange 3, Technical Grade, TRC

Artikelnummer. 30328449
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30328449 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30328449 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18988310G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Acid Orange 3
CAS 6373-74-6
Molekylær formel C18 H13 N4 O7 S . Na
Mængde 10 g
Synonym Benzenesulfonic acid, 2-anilino-5-(2,4-dinitroanilino)-, monosodium salt (8CI), Benzenesulfonic acid, 5-[(2,4-dinitrophenyl)amino]-2-(phenylamino)-, monosodium salt (9CI), C.I. Acid Orange 3 (7CI), Acid Fast Yellow AG, Acid Fast Yellow E 5R, Acid Fast Yellow ES, Acid Leather Light Brown G, Acid Orange 3, Acid Orange E-3R, Acid Orange No. 3, Acid Orange R, Acid Yellow AE, Acid Yellow Brown 3RL, Acid Yellow E, Airedale Yellow E, Amido Yellow E, Amido Yellow EA, Amido Yellow EA-CF, Amido Yellow EN, Anadurm Orange A-E, Anthralan Yellow RRT, Apollo Nylon Fast Yellow Brown E-L, Best Acid Yellow Brown GT, C.I. 10385, Colocid Yellow Brown E3R, Covalene Orange E, Derma Fur Yellow RT, Derma Yellow P, Dinacid Fast Yellow BRD, Duasyn Acid Yellow RRT 01, Dyacid Yellow A, Dynacid Orange 3D, Erio Fast Yellow AE, Erio Fast Yellow AEN, Erionyl Yellow E-AEN, Fast Light Yellow E, Fenalan Yellow E, Formo Acid Orange III, Kemacid Fur Yellow RT, Kenamide Yellow GAE, Kiton Fast Yellow A, Light Fast Yellow ES, Lissamine Fast Yellow AE, Lissamine Fast Yellow AES, Lissamine Yellow AE, Lissamine Yellow AE 110, Multacid Yellow 3R, Multicuer Brown MPH, Naphthazine Orange E, Nylochrom Yellow 3R, Nylomine Acid Yellow B-RD, Rifa Acid Fast Yellow Brown E-L, Rifa Leather Brown PS, Sodium 4-(2,4-dinitroanilino)diphenylamine-2-sulfonate, Superian Yellow R, Tectilon Orange 3GT, Tertracid Light Yellow 2R, Triacid Light Yellow AE, Vicoacid Orange 3, Vondacid Fast Yellow AE, Xylene Fast Yellow ES, Yellow for Furs R
InChI formel InChI=1S/C18H14N4O7S.Na/c23-21(24)14-7-9-15(17(11-14)22(25)26)20-13-6-8-16(18(10-13)30(27,28)29)19-12-4-2-1-3-5-12;/h1-11,19-20H,(H,27,28,29);/q;+1/p-1
SMIL [Na+].[O-][N+](=O)c1ccc(Nc2ccc(Nc3ccccc3)c(c2)S(=O)(=O)[O-])c(c1)[N+](=O)[O-]
IUPAC navn sodium;2-anilino-5-(2,4-dinitroanilino)benzenesulfonate
Molekylvægt (g/mol) 452.3732
Formel vægt 452.0403
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.