Få mere at vide
Acid Orange 24 (>90%), TRC
Artikelnummer. 30134577
Skift visning
Mængde:
1 mg
5 mg
10 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30134577 5 mg
18023540 1 mg
18052920 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
Artikelnummer. 30134577 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1899555MG
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternative produkter
Regulatorisk: Dette produkt falder ind under Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. Hvis det er relevant, skal kunden fremskaffe en licens før levering.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 1320-07-6
Molekylær formel C20 H17 N4 O5 S . Na
Molekylvægt (g/mol) 448.43
Synonym Benzenesulfonic acid, 4-[[3-[(dimethylphenyl)azo]-2,4-dihydroxyphenyl]azo]-, monosodium salt (9CI), C.I. Acid Orange 24 (7CI), C.I. Acid Orange 24, monosodium salt (8CI), Acid Orange 24, Albion Leather Brown FH, Brown 201, Brown No. 201, C Ext. Brown 4, C.I. 20170, D & C Brown 1, D and C Brown No. 1, D&C Brown No. 1, Fabracid Brown S-B, Japan Brown 201, Japan Brown No. 201, Leather Brown 2G, Leather Brown RGN, Multicuer Brown MNO, Ratna Acid Fast Brown 2BB, Resorcin Brown, Resorcin Brown G, Resorcin Brown R, Resorcine Brown B, Resorcine Brown G, Resorcine Brown R;Sodium 4-((2,4-Dimethylphenyl)diazenyl)-2,4-dihydroxyphenyl)diazenyl)benzenesulfonate, Sodium,4-[(2Z)-2-[(5E)-5-[(2,4-dimethylphenyl)hydrazinylidene]-4,6-dioxocyclohex-2-en-1-ylidene]hydrazinyl]benzenesulfonate, C.I. Acid Orange 24, Monosodium Salt, Albion Leather Brown FH, Brown 201, Brown No. 201, C Ext. Brown 4, C.I. 20170, D & C Brown 1, D and C Brown No. 1, D&C Brown No. 1, Fabracid Brown S-B, Japan Brown 201, Japan Brown No. 201, Leather Brown 2G, Leather Brown RGN, Multicuer Brown MNO, Ratna Acid Fast Brown 2BB, Resorcin Brown, Resorcin Brown G, Resorcin Brown R, Resorcine Brown B, Resorcine Brown G, Resorcine Brown R
IUPAC navn sodium;4-[(E)-[3-[(Z)-(2,4-dimethylphenyl)azo]-2,4-dihydroxy-phenyl]azo]benzenesulfonate
SMIL [Na+].Cc1ccc(N=Nc2c(O)ccc(N=Nc3ccc(cc3)S(=O)(=O)[O-])c2O)c(C)c1
Tekniske data
Kemisk navn eller materiale Acid Orange 24
Analyt- eller komponentnavne Acid Orange 24
Forsendelsestilstand Room Temperature
InChI formel InChI=1S/C20H18N4O5S.Na/c1-12-3-8-16(13(2)11-12)22-24-19-18(25)10-9-17(20(19)26)23-21-14-4-6-15(7-5-14)30(27,28)29;/h3-11,25-26H,1-2H3,(H,27,28,29);/q;+1/p-1/b23-21+,24-22-;
Løsningstype Neat
Industry Type Pharmaceutical
Mængde 5 mg
Formel vægt 448.0817