Få mere at vide
Acid Green 9 (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acid Green 9 |
| CAS | 4857-81-2 |
| Molekylær formel | C37H34ClN2NaO6S2 |
| Mængde | 2.5 g |
| Synonym | Benzenemethanaminium, N-[4-[(2-chlorophenyl)[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1- ylidene]-N-ethyl-3-sulfo-, hydroxide, inner salt, sodium salt, Benzenemethanaminium, N-[4-[(2-chlorophenyl)[4-[ethyl[(3- sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-, inner salt, sodium salt (9CI), Brilliant Milling Green B (6CI), C.I. Acid Green 9 (7CI), C.I. Acid Green 9, monosodium salt (8CI), 4-[[4-(N-Ethyl-m-sulfobenzylamino)phenyl](o-chlorophenyl)methylene]-1-(N-ethyl-Nm-sulfonium-benzyl-∆2,5-cyclohexadienimine)monosodium salt, Acid Brilliant Green 6B, Acid Brilliant Green B, Acid Green 5B, Acid Green 6B, Acid Green 9, Acid Green GB, Acid Leather Green KGB, Acilan Brilliant Green 6B, Airedale Green B, Aizen Acid Brilliant Green B, Aizen Brilliant Acid Green 6BH, Aizen Brilliant Acid Green 6BH Extra, Aizen Brilliant Milling Green B, Alizarin Green, Amacid Milling Green B, Bezanyl Green F 2B, Brilliant Acid Green 6B, Brilliant Acid Green B, Brilliant Milling Green, Brilliant Milling Green NS, Bucacid Fast Green B, C.I. 42100, Calcocid Milling Green 6B, Cetil Brilliant Green 6B, Colocid Green 6B, Dinacid Milling Green B, Disulphine Green B, Erio Viridine B, Eriogreen, Fast Acid Green B, Fenazo Green XG, Folan Green B, Hidacid Fast Green B, Indigo Green B, Kiton Green B, Leather Green 6B, Lerui Acid Brilliant Green 6B, Merantine Green B, Milling Brilliant Green B, Milling Green, Mitsui Acid Brilliant Milling Green BM 100, Mitsui Brilliant Milling Green BX, NSC 4319, NSC 51540, NSC 8683, Neptune Green SGXA, Night Green A, Sandolan Brilliant Green E-6B, Sumitomo Brilliant Milling Green B, Triacid Green 6B, Vicoacid Green 9, Xylene Fast Green 6B, Xylene Milling Green 6B |
| InChI formel | InChI=1S/C37H35ClN2O6S2.Na/c1-3-39(25-27-9-7-11-33(23-27)47(41,42)43)31-19-15-29(16-20-31)37(35-13-5-6-14-36(35)38)30-17-21-32(22-18-30)40(4-2)26-28-10-8-12-34(24-28)48(44,45)46;/h5-24H,3-4,25-26H2,1-2H3,(H-,41,42,43,44,45,46);/q;+1/p-1 |
| SMIL | CCN(C1=CC=C(/C(C2=CC=CC=C2Cl)=C3C=C/C(C=C/3)=[N+](CC4=CC(S(=O)([O-])=O)=CC=C4)\CC)C=C1)CC5=CC(S(=O)(O[Na])=O)=CC=C5 |
| IUPAC navn | sodium;3-[[4-[(2-chlorophenyl)-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate |
| Molekylvægt (g/mol) | 725.25 |
| Formel vægt | 724.1445 |