missing translation for 'onlineSavingsMsg'
Få mere at vide

Acid Green 9 (Technical Grade), TRC

Artikelnummer. 30309766
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30309766 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30309766 Leverandør Toronto Research Chemicals Leverandørnr. TRCA189908250MG

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Acid Green 9
CAS 4857-81-2
Molekylær formel C37H34ClN2NaO6S2
Mængde 250 mg
Synonym Benzenemethanaminium, N-[4-[(2-chlorophenyl)[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1- ylidene]-N-ethyl-3-sulfo-, hydroxide, inner salt, sodium salt, Benzenemethanaminium, N-[4-[(2-chlorophenyl)[4-[ethyl[(3- sulfophenyl)methyl]amino]phenyl]methylene]-2,5-cyclohexadien-1-ylidene]-N-ethyl-3-sulfo-, inner salt, sodium salt (9CI), Brilliant Milling Green B (6CI), C.I. Acid Green 9 (7CI), C.I. Acid Green 9, monosodium salt (8CI), 4-[[4-(N-Ethyl-m-sulfobenzylamino)phenyl](o-chlorophenyl)methylene]-1-(N-ethyl-Nm-sulfonium-benzyl-∆2,5-cyclohexadienimine)monosodium salt, Acid Brilliant Green 6B, Acid Brilliant Green B, Acid Green 5B, Acid Green 6B, Acid Green 9, Acid Green GB, Acid Leather Green KGB, Acilan Brilliant Green 6B, Airedale Green B, Aizen Acid Brilliant Green B, Aizen Brilliant Acid Green 6BH, Aizen Brilliant Acid Green 6BH Extra, Aizen Brilliant Milling Green B, Alizarin Green, Amacid Milling Green B, Bezanyl Green F 2B, Brilliant Acid Green 6B, Brilliant Acid Green B, Brilliant Milling Green, Brilliant Milling Green NS, Bucacid Fast Green B, C.I. 42100, Calcocid Milling Green 6B, Cetil Brilliant Green 6B, Colocid Green 6B, Dinacid Milling Green B, Disulphine Green B, Erio Viridine B, Eriogreen, Fast Acid Green B, Fenazo Green XG, Folan Green B, Hidacid Fast Green B, Indigo Green B, Kiton Green B, Leather Green 6B, Lerui Acid Brilliant Green 6B, Merantine Green B, Milling Brilliant Green B, Milling Green, Mitsui Acid Brilliant Milling Green BM 100, Mitsui Brilliant Milling Green BX, NSC 4319, NSC 51540, NSC 8683, Neptune Green SGXA, Night Green A, Sandolan Brilliant Green E-6B, Sumitomo Brilliant Milling Green B, Triacid Green 6B, Vicoacid Green 9, Xylene Fast Green 6B, Xylene Milling Green 6B
InChI formel InChI=1S/C37H35ClN2O6S2.Na/c1-3-39(25-27-9-7-11-33(23-27)47(41,42)43)31-19-15-29(16-20-31)37(35-13-5-6-14-36(35)38)30-17-21-32(22-18-30)40(4-2)26-28-10-8-12-34(24-28)48(44,45)46;/h5-24H,3-4,25-26H2,1-2H3,(H-,41,42,43,44,45,46);/q;+1/p-1
SMIL CCN(C1=CC=C(/C(C2=CC=CC=C2Cl)=C3C=C/C(C=C/3)=[N+](CC4=CC(S(=O)([O-])=O)=CC=C4)\CC)C=C1)CC5=CC(S(=O)(O[Na])=O)=CC=C5
IUPAC navn sodium;3-[[4-[(2-chlorophenyl)-[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]methyl]-N-ethylanilino]methyl]benzenesulfonate
Molekylvægt (g/mol) 725.25
Formel vægt 724.1445
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.