Få mere at vide
Acid Green 50 (Technical Grade), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acid Green 50 |
| CAS | 3087-16-9 |
| Molekylær formel | C27 H25 N2 O7 S2 . Na |
| Mængde | 1 g |
| Synonym | C.I. Acid Green 50 (7CI), C.I. Acid Green 50, monosodium salt (8CI), Lissamine Green BN (6CI), Methanaminium, N-[4-[[4-(dimethylamino)phenyl](2-hydroxy-3,6-disulfo-1-naphthalenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, hydroxide, inner salt, monosodium salt, Methanaminium, N-[4-[[4-(dimethylamino)phenyl](2-hydroxy-3,6-disulfo-1-naphthalenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, inner salt, monosodium salt (9CI), 12078 Green, Acid Brilliant Green BS, Acid Green 50, Acid Green BS, Acid Green RS, Acid Leather Green S, Acidal Wool Green BS, Acilan Green BS, Amacid Wool Green S, Ariavit Green S, Brilliant Acid Green BS, Bucacid Wool Green, C.I. 44090, C.I. Food Green 4, Calcocid Green S, Calcocid Green SB, Dycosacid Green BS, E 142, Edicol Supra Green B, Edicol Supra Green BS, Erio Green S, Eurocert Green S 312524, Green H 002, Green S, Green S 44090, Hexacol Green S, Hidacid Wool Green, Kiton Green S, Lissamine Green B, Naphthazine Green S, Pharmacid Green S, Sumitomo Wool Green S, Unitertracid Green BS, Vondacid Green S, Water Green 176558, Water Green SX, Wool Green BS, Wool Green BSNA, Wool Green MS, Wool Green S, Wool Green SF, Wool Green SG |
| InChI formel | InChI=1S/C27H26N2O7S2.Na/c1-28(2)20-9-5-17(6-10-20)25(18-7-11-21(12-8-18)29(3)4)26-23-14-13-22(37(31,32)33)15-19(23)16-24(27(26)30)38(34,35)36;/h5-16H,1-4H3,(H2,31,32,33,34,35,36);/q;+1/p-1 |
| SMIL | [Na+].CN(C)c1ccc(cc1)C(=C2C=CC(=[N+](C)C)C=C2)c3c(O)c(cc4cc(ccc34)S(=O)(=O)[O-])S(=O)(=O)[O-] |
| IUPAC navn | sodium;4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-3-hydroxynaphthalene-2,7-disulfonate |
| Molekylvægt (g/mol) | 576.62 |
| Formel vægt | 576.1001 |