missing translation for 'onlineSavingsMsg'
Få mere at vide

Acid Green 50 (Technical Grade), TRC

Artikelnummer. 30312543
Skift visning
Klik for at se tilgængelige muligheder
Mængde:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30312543 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 30312543 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18989525G

Please to purchase this item. Need a web account? Register with us today!

Denne vare kan ikke returneres. Se returpolicy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Tekniske data

Kemisk navn eller materiale Acid Green 50
CAS 3087-16-9
Molekylær formel C27 H25 N2 O7 S2 . Na
Mængde 25 g
Synonym C.I. Acid Green 50 (7CI), C.I. Acid Green 50, monosodium salt (8CI), Lissamine Green BN (6CI), Methanaminium, N-[4-[[4-(dimethylamino)phenyl](2-hydroxy-3,6-disulfo-1-naphthalenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, hydroxide, inner salt, monosodium salt, Methanaminium, N-[4-[[4-(dimethylamino)phenyl](2-hydroxy-3,6-disulfo-1-naphthalenyl)methylene]-2,5-cyclohexadien-1-ylidene]-N-methyl-, inner salt, monosodium salt (9CI), 12078 Green, Acid Brilliant Green BS, Acid Green 50, Acid Green BS, Acid Green RS, Acid Leather Green S, Acidal Wool Green BS, Acilan Green BS, Amacid Wool Green S, Ariavit Green S, Brilliant Acid Green BS, Bucacid Wool Green, C.I. 44090, C.I. Food Green 4, Calcocid Green S, Calcocid Green SB, Dycosacid Green BS, E 142, Edicol Supra Green B, Edicol Supra Green BS, Erio Green S, Eurocert Green S 312524, Green H 002, Green S, Green S 44090, Hexacol Green S, Hidacid Wool Green, Kiton Green S, Lissamine Green B, Naphthazine Green S, Pharmacid Green S, Sumitomo Wool Green S, Unitertracid Green BS, Vondacid Green S, Water Green 176558, Water Green SX, Wool Green BS, Wool Green BSNA, Wool Green MS, Wool Green S, Wool Green SF, Wool Green SG
InChI formel InChI=1S/C27H26N2O7S2.Na/c1-28(2)20-9-5-17(6-10-20)25(18-7-11-21(12-8-18)29(3)4)26-23-14-13-22(37(31,32)33)15-19(23)16-24(27(26)30)38(34,35)36;/h5-16H,1-4H3,(H2,31,32,33,34,35,36);/q;+1/p-1
SMIL [Na+].CN(C)c1ccc(cc1)C(=C2C=CC(=[N+](C)C)C=C2)c3c(O)c(cc4cc(ccc34)S(=O)(=O)[O-])S(=O)(=O)[O-]
IUPAC navn sodium;4-[[4-(dimethylamino)phenyl]-(4-dimethylazaniumylidenecyclohexa-2,5-dien-1-ylidene)methyl]-3-hydroxynaphthalene-2,7-disulfonate
Molekylvægt (g/mol) 576.62
Formel vægt 576.1001
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.