Få mere at vide
Acid Green 3 (~50%), TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acid Green 3 |
| CAS | 4680-78-8 |
| Molekylær formel | C37H35N2NaO6S2 |
| Mængde | 25 g |
| Synonym | N-Ethyl-N-[4-[[4-[ethyl[(3-sulfophenyl)methyl]amino]phenyl]phenylmethylene]-2,5-cyclohexadien-1-ylidene]-3-sulfo-benzenemethanaminium Inner Salt Sodium Salt, C.I. Acid Green 3, C.I. Acid Green 3 Sodium Salt, 4-[4-(N-Ethyl-p-sulfobenzylamino)diphenylmethylene]-[1-(N-ethyl-N-p-sulfoniumbenzyl)-delta2,5-cyclohexadienimine] Monosodium Salt, A.F. Green No. 1, Acid Green, Acid Green 2G, Acid Green B, Acid Green G, Acid Green G (Polish), Acid Green S, Acid Leather Green 3G, Acid Leather Green F, Acid green 3, Acidal Green G, Acilan Green B, Amacid Green B, Brilliant Green 3EMBL, Bucacid Guinea Green BA, C.I. 42085C.I. Food Green 1, Calcocid Green G, FD and C Green No. 1, FD&C Green No. 1, Fenazo Green L, Guinea Green B, Guinea Green BA, Guinea green, Hidacid Emerald Green, Hispacid Green GB, Japan Green 1, Japan Green 402, Japan Green No. 1Japan Green No. 402, Kiton Green F, Kiton Green FC, Leather Green B, Merantine Green G, NSC 5016, NSC 8684, Naphthalene Green G, Naphthalene Lake Green G, Naphthalene Leather Green G, Neran Brilliant Green G, Pontacyl Green B, Pontacyl Green BL, Sulpho Green 2B, Triacid Green G, Vondacid Green L |
| InChI formel | InChI=1S/C37H36N2O6S2.Na/c1-3-38(26-28-10-8-14-35(24-28)46(40,41)42)33-20-16-31(17-21-33)37(30-12-6-5-7-13-30)32-18-22-34(23-19-32)39(4-2)27-29-11-9-15-36(25-29)47(43,44)45;/h5-25H,3-4,26-27H2,1-2H3,(H-,40,41,42,43,44,45);/q;+1/p-1 |
| SMIL | [Na+].CCN(Cc1cccc(c1)S(=O)(=O)[O-])c2ccc(cc2)C(=C3C=CC(=[N+](CC)Cc4cccc(c4)S(=O)(=O)[O-])C=C3)c5ccccc5 |
| IUPAC navn | sodium;3-[[N-ethyl-4-[[4-[ethyl-[(3-sulfonatophenyl)methyl]azaniumylidene]cyclohexa-2,5-dien-1-ylidene]-phenylmethyl]anilino]methyl]benzenesulfonate |
| Molekylvægt (g/mol) | 690.8 |
| Formel vægt | 690.1834 |