Få mere at vide
Acid Blue 25, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acid Blue 25 |
| CAS | 6408-78-2 |
| Molekylær formel | C20H13N2NaO5S |
| Mængde | 5 g |
| Synonym | 1-Amino-4-anilinoanthraquinone-2-sulfonic acid sodium salt, Acid Alizarine Blue A, Acid Blue 2AL241, Ambinyl Blue LC, Apollo Nylon Fast Blue AApollo Nylon Fast Blue A 2G, Atul Acid Blue ARB, Best Acid Blue AGL, Best Acid Sky Blue B, C.I. Acid Blue 25 sodium salt, C.I. Acid Blue 77, Colocid Fast Blue AConacid Blue BE, Concorde Acid Blue A-GL, Erio Anthracene Brilliant Blue GRLErio Fast Blue GRL, Fenazo Light Blue AT, Formo Acid Blue 25, Triacid Light Blue A, Vicoacid Blue 25, Wofacid Blue A, Wofasol Fast Blue L |
| InChI formel | InChI=1S/C20H14N2O5S.Na/c21-18-15(28(25,26)27)10-14(22-11-6-2-1-3-7-11)16-17(18)20(24)13-9-5-4-8-12(13)19(16)23;/h1-10,22H,21H2,(H,25,26,27);/q;+1/p-1 |
| SMIL | [Na+].Nc1c2C(=O)c3ccccc3C(=O)c2c(Nc4ccccc4)cc1S(=O)(=O)[O-] |
| IUPAC navn | sodium;1-amino-4-anilino-9,10-dioxoanthracene-2-sulfonate |
| Molekylvægt (g/mol) | 416.38 |
| Formel vægt | 416.0443 |