Få mere at vide
Acetylsalicylic Acid EP Impurity B (Salicylic Acid USP Related Compound B), Mikromol™
Artikelnummer. 30459304
Skift visning
Mængde:
25 mg
100 mg
4 x 25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30459304 4 x 25 mg
30052392 25 mg
30050564 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30459304 Leverandør Mikromol Leverandørnr. MM0045.0200254AMP
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
Acetylsalicylic Acid EP Impurity B (Salicylic Acid USP Related Compound B), Mikromol
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale 4-Hydroxyisophthalic Acid
CAS 636-46-4
Analyt- eller komponentnavne 4-Hydroxyisophthalic Acid
Forsendelsestilstand Room Temperature
Molekylær formel C8 H6 O5
Certificeringer/compliance ISO 17025
Synonym Acetylsalicylic Acid Imp. B (EP), Salicylic Acid Imp. B (EP), Salicylic Acid USP Related Compound B, Salicylic Acid USP RC B, 4-Hydroxybenzene-1,3-dicarboxylic Acid, 4-Hydroxyisophthalic Acid, 1,3-Benzenedicarboxylic acid, 4-hydroxy-, Isophthalic acid, 4-hydroxy- (6CI,7CI,8CI), 4-Hydroxy-1,3-benzenedicarboxylic acid, 4-Hydroxy-m-phthalic acid, Eupirina, NSC 2445, Ph Eur Salicylic Acid Impurity B, USP Salicylic Acid Related Compound B, Salicylic Acid Related Compound B, Acetylsalicylic Acid Impurity B, Salicylic Acid Impurity B
InChI formel InChI=1S/C8H6O5/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3,9H,(H,10,11)(H,12,13)
Løsningstype Neat
SMIL OC(=O)c1ccc(O)c(c1)C(=O)O
IUPAC navn 4-hydroxybenzene-1,3-dicarboxylic acid
Molekylvægt (g/mol) 182.13
Mængde 4 x 25 mg
Formel vægt 182.0215 g/mol