Få mere at vide
Acetylsalicylic Acid-d4, TRC
Artikelnummer. 30320074
Skift visning
Mængde:
25 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30320074 50 mg
30318087 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320074 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18778250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylsalicylic Acid-d4
CAS 97781-16-3
Renhedsgrad noter HPLC
Molekylær formel C9 2H4 H4 O4
Mængde 50 mg
Synonym Benzoic-2,3,4,5-d4 acid, 6-(acetyloxy)- (9CI), Acetylsalicylic acid-d4, 6-(Acetyloxy)benzoic-2,3,4,5-d4 acid, 2-Acetyloxybenzoic-3,4,5,6-d4 acid, Acetylsalicylic Acid-D4 (phenyl-D4), Aspirin-d4
InChI formel InChI=1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C(=O)O)c(OC(=O)C)c1[2H]
IUPAC navn 2-acetyloxy-3,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 184.18
Formel vægt 184.0674
Procent renhed >95