Få mere at vide
Acetylsalicylic Acid-d4, TRC
Artikelnummer. 30314212
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30314212 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314212 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1877825MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylsalicylic Acid-d4
CAS 97781-16-3
Renhedsgrad noter HPLC
Molekylær formel C9 2H4 H4 O4
Mængde 5 mg
Synonym Benzoic-2,3,4,5-d4 acid, 6-(acetyloxy)- (9CI), Acetylsalicylic acid-d4, 6-(Acetyloxy)benzoic-2,3,4,5-d4 acid, 2-Acetyloxybenzoic-3,4,5,6-d4 acid, Acetylsalicylic Acid-D4 (phenyl-D4), Aspirin-d4
InChI formel InChI=1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)/i2D,3D,4D,5D
SMIL [2H]c1c([2H])c([2H])c(C(=O)O)c(OC(=O)C)c1[2H]
IUPAC navn 2-acetyloxy-3,4,5,6-tetradeuteriobenzoic acid
Molekylvægt (g/mol) 184.18
Formel vægt 184.0674
Procent renhed >95