Få mere at vide
Acetylsalicylic Acid-d3, TRC
Artikelnummer. 30308800
Skift visning
Mængde:
50 mg
Artikelnummer. Mængde
30308800 50 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30308800 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18709250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylsalicylic Acid-d3
CAS 921943-73-9
Molekylær formel C9 2H3 H5 O4
Mængde 50 mg
Synonym Benzoic acid, 2-(acetyl-2,2,2-d3-oxy)- (ACI), 2-(Acetyl-2,2,2-d3-oxy)benzoic acid (ACI), 2-(Acetyloxy-d3)benzoic acid, Acetylsalicylic acid-d3, Acetylsalicylic Acid-D3 (acetyl-D3), Aspirin-d3
InChI formel InChI=1S/C9H8O4/c1-6(10)13-8-5-3-2-4-7(8)9(11)12/h2-5H,1H3,(H,11,12)/i1D3
SMIL [2H]C([2H])([2H])C(=O)Oc1ccccc1C(=O)O
IUPAC navn 2-(2,2,2-trideuterioacetyl)oxybenzoic acid
Molekylvægt (g/mol) 183.18
Formel vægt 183.0611