Få mere at vide
Acetylhistamine, TRC
Artikelnummer. 30326147
Skift visning
Mængde:
5 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326147 50 mg
30312321 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30326147 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17911050MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylhistamine
CAS 673-49-4
Molekylær formel C7H11N3O
Mængde 50 mg
Synonym N-[2-(1H-Imidazol-5-yl)ethyl]acetamide, N-[2-imidazol-4(or 5)-ylethyl]acetamide
InChI formel InChI=1S/C7H11N3O/c1-6(11)9-3-2-7-4-8-5-10-7/h4-5H,2-3H2,1H3,(H,8,10)(H,9,11)
SMIL CC(NCCC1=CN=CN1)=O
IUPAC navn N-[2-(1H-imidazol-5-yl)ethyl]acetamide
Molekylvægt (g/mol) 153.18
Formel vægt 153.0902