Få mere at vide
Acetylene-linker-Val-Cit-PABC-MMAE, MedChemExpress
Artikelnummer. 16450388
Encompass_Preferred
Skift visning
Mængde:
1 mg
5 mg
10 mg
Pakningsstørrelse:
10mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde unitSize
16450388 5 mg -
16411879 1 mg -
16407928 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 16450388 Leverandør Medchem Express Leverandørnr. HY19812
MedChemExpress Acetylene-linker-Val-Cit-PABC-MMAE (LCB14-0602) consists the ADCs linker (Acetylene-linker-Val-Cit-PABC) and potent tubulin inhibitor (MMAE). Acetylene-linker-Val-Cit-PABC-MMAE (LCB14-0602) is a drug-linker conjugate for ADC.
Acetylene-linker-Val-Cit-PABC-MMAE (LCB14-0602) consists the ADCs linker (Acetylene-linker-Val-Cit-PABC) and potent tubulin inhibitor (MMAE). Acetylene-linker-Val-Cit-PABC-MMAE (LCB14-0602) is a drug-linker conjugate for ADC.
Tekniske data
Kemisk navn eller materiale Acetylene-linker-Val-Cit-PABC-MMAE
CAS 1411977-95-1
Farve White
Renhedsgrad noter Research
Molekylær formel C67H106N10O16
Synonym LCB14-0602
SMIL O=C([C@H](CCCNC(N)=O)NC([C@H](C(C)C)NC(COCCOCCOCC#C)=O)=O)NC1=CC=C(COC(N(C)[C@@H](C(C)C)C(N[C@@H](C(C)C)C(N([C@H]([C@@H](CC(N2[C@H]([C@H](OC)[C@H](C(N[C@@H]([C@@H](O)C3=CC=CC=C3)C)=O)C)CCC2)=O)OC)[C@@H](C)CC)C)=O)=O)=O)C=C1
Molekylvægt (g/mol) 1307.62
Mængde 5 mg
Formel vægt 1307.62
Procent renhed 95.08%
Grad Research
Fysisk form Solid
Til brug med (applikation) Cancer-programmed cell death