Få mere at vide
Acetylcholine-d9 Chloride (N,N,N-trimethyl-d9), CDN
Artikelnummer. 30323434
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30323434 0.05 g
30323886 0.01 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30323434 Leverandør CDN Leverandørnr. CDND25590.05G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylcholine-d9 Chloride
CAS 344298-95-9
Molekylær formel C7 2H9 H7 N O2 . Cl
Mængde 0.05 g
Synonym Ethanaminium, 2-(acetyloxy)-N,N,N-tri(methyl-d3)-, chloride (9 CI), Acetylcholine-d9 Chloride, Acetylcholine-D9 Chloride (N,N,N-trimethyl D9)
InChI formel InChI=1 S/C7H16NO2.ClH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1 H/q+1;/p-1/i2D3,3D3,4D3;
SMIL [Cl-].[2 H]C([2 H])([2 H])[N+](CCOC(=O)C)(C([2 H])([2 H])[2 H])C([2 H])([2 H])[2 H]
IUPAC navn 2-acetyloxyethyl-tris(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 190.72
Formel vægt 190.1434 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity