Få mere at vide
Acetylcholine-d9 Bromide, TRC
Artikelnummer. 30316384
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30316384 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30316384 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1720525MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylcholine-d9 Bromide
CAS 93449-32-2
Molekylær formel C7 2H9 H7 N O2 . Br
Mængde 5 mg
Synonym Ethanaminium, 2-(acetyloxy)-N,N,N-tri(methyl-d3)-, bromide (9CI), Acetylcholine-d9 Bromide, Acetylcholine-D9 Bromide (N,N,N-trimethyl D9)
InChI formel InChI=1S/C7H16NO2.BrH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1/i2D3,3D3,4D3;
SMIL [Br-].[2H]C([2H])([2H])[N+](CCOC(=O)C)(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn 2-acetyloxyethyl-tris(trideuteriomethyl)azanium;bromide
Molekylvægt (g/mol) 235.17
Formel vægt 234.0929