Få mere at vide
Acetylcholine-d4 Chloride, TRC
Artikelnummer. 30322088
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30322088 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30322088 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17206310MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylcholine-d4 Chloride
CAS 344298-94-8
Molekylær formel C7 D4 H12 N O2 . Cl
Mængde 10 mg
Synonym Ethan-1,1,2,2-d4-aminium, 2-(acetyloxy)-N,N,N-trimethyl-, chloride (1:1) (ACI), 2-(Acetyloxy)-N,N,N-trimethylethan-1,1,2,2-d4-aminium chloride (1:1) (ACI), Ethan-1,1,2,2-d4-aminium, 2-(acetyloxy)-N,N,N-trimethyl-, chloride (9CI), Acetylcholine-1,1,2,2-d4 Chloride, Acetylcholine-d4 Chloride, Acetylcholine-1,1,2,2-D4 Chloride
InChI formel InChI=1S/C7H16NO2.ClH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1/i5D2,6D2;
SMIL [Cl-].[2H]C([2H])(OC(=O)C)C([2H])([2H])[N+](C)(C)C
IUPAC navn (2-acetyloxy-1,1,2,2-tetradeuterioethyl)-trimethylazanium;chloride
Molekylvægt (g/mol) 185.685
Formel vægt 185.112