Få mere at vide
Acetylcholine-d16 Bromide, TRC
Artikelnummer. 30320150
Skift visning
Mængde:
5 mg
Artikelnummer. Mængde
30320150 5 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30320150 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1720645MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylcholine-D16 Bromide
CAS 347841-43-4
Molekylær formel C7 D16 N O2 . Br
Mængde 5 mg
Synonym Ethan-1,1,2,2-d4-aminium, 2-(acetyl-d3-oxy)-N,N,N-tri(methyl-d3)-, bromide (9CI), 2-(Acetyl-d3-oxy)-N,N,N-tri(methyl-d3)ethan-1,1,2,2-d4-aminium bromide (1:1), Acetylcholine Bromide-d16, Acetylcholine-d16 Bromide, Acetylcholine-D16 Bromide
InChI formel InChI=1S/C7H16NO2.BrH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1/i1D3,2D3,3D3,4D3,5D2,6D2;
SMIL [Br-].[2H]C([2H])([2H])C(=O)OC([2H])([2H])C([2H])([2H])[N+](C([2H])([2H])[2H])(C([2H])([2H])[2H])C([2H])([2H])[2H]
IUPAC navn [1,1,2,2-tetradeuterio-2-(2,2,2-trideuterioacetyl)oxyethyl]-tris(trideuteriomethyl)azanium;bromide
Molekylvægt (g/mol) 242.21
Formel vægt 241.1369