Få mere at vide
Acetylcholine-1,1,2,2-d4 Chloride, CDN
Artikelnummer. 30311130
Skift visning
Mængde:
0.01 g
0.05 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30311130 0.01 g
30310645 0.05 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30311130 Leverandør CDN Leverandørnr. CDND25580.01G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylcholine-d4 Chloride
CAS 344298-94-8
Molekylær formel C7 D4 H12 N O2 . Cl
Mængde 0.01 g
Synonym Ethan-1,1,2,2-d4-aminium, 2-(acetyloxy)-N,N,N-trimethyl-, chloride (1:1) (ACI), 2-(Acetyloxy)-N,N,N-trimethylethan-1,1,2,2-d4-aminium chloride (1:1) (ACI), Ethan-1,1,2,2-d4-aminium, 2-(acetyloxy)-N,N,N-trimethyl-, chloride (9 CI), Acetylcholine-1,1,2,2-d4 Chloride, Acetylcholine-d4 Chloride, Acetylcholine-1,1,2,2-D4 Chloride
InChI formel InChI=1 S/C7H16NO2.ClH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1 H/q+1;/p-1/i5D2,6D2;
SMIL [Cl-].[2 H]C([2 H])(OC(=O)C)C([2 H])([2 H])[N+](C)(C)C
IUPAC navn (2-acetyloxy-1,1,2,2-tetradeuterioethyl)-trimethylazanium;chloride
Molekylvægt (g/mol) 185.685
Formel vægt 185.112 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity