Få mere at vide
Acetylcholine-1,1,2,2-d4 Bromide, TRC
Artikelnummer. 30318463
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30318463 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30318463 Leverandør Toronto Research Chemicals Leverandørnr. TRCA17207210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetylcholine-1,1,2,2-D4 Bromide
CAS 93449-31-1
Molekylær formel C7 D4 H12 N O2 . Br
Mængde 10 mg
Synonym Ethan-1,1,2,2-d4-aminium, 2-(acetyloxy)-N,N,N-trimethyl-, bromide (9CI), 2-(Acetyloxy)-N,N,N-trimethylethan-1,1,2,2-d4-aminium bromide (1:1), Acetylcholine-1,1,2,2-d4 Bromide, Acetylcholine-d4 Bromide, Acetylcholine-1,1,2,2-D4 Bromide
InChI formel InChI=1S/C7H16NO2.BrH/c1-7(9)10-6-5-8(2,3)4;/h5-6H2,1-4H3;1H/q+1;/p-1/i5D2,6D2;
SMIL [Br-].[2H]C([2H])(OC(=O)C)C([2H])([2H])[N+](C)(C)C
IUPAC navn (2-acetyloxy-1,1,2,2-tetradeuterioethyl)-trimethylazanium;bromide
Molekylvægt (g/mol) 230.1361
Formel vægt 229.0615