Få mere at vide
Acetyl Vitamin B12, TRC
Artikelnummer. 17968994
Skift visning
Mængde:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
17968994 10 mg
17977104 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 17968994 Leverandør Toronto Research Chemicals Leverandørnr. TRCA18802810MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 855207-36-2
Molekylær formel C65 H90 Co N14 O15 P
Synonym Coβ-cyano-O5R-acetylcobalamin
Tekniske data
Kemisk navn eller materiale Acetyl Vitamin B12
Mængde 10 mg
Forsendelsestilstand Room Temperature
Formel vægt 1396.578
Molekylvægt (g/mol) 1397.4, 1397.43
SMIL [Co++].[C-]#N.[H]C12[N-]\C(=C(C)/C3=N/C(=C\C4=N\C(=C(C)/C5=NC1(C)C(C)(CC(N)=O)C5CCC(N)=O)\C(C)(CC(N)=O)C4CCC(N)=O)/C(C)(C)C3CCC(N)=O)C(C)(CCC(=O)NCC(C)OP([O-])(=O)O[C@@H]1[C@@H](COC(C)=O)O[C@@H]([C@@H]1O)n1c[nH]c3cc(C)c(C)cc13)C2CC(N)=O