Få mere at vide
Acetyl Propionate, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acetic Propionic Anhydride |
| CAS | 13080-96-1 |
| Molekylær formel | C5 H8 O3 |
| Mængde | 250 mg |
| Synonym | Propanoic acid, anhydride with acetic acid, Propionic acid, anhydride with AcOH (6CI,7CI), Propionic acid, anhydride with acetic acid (8CI), Acetic acid, anhydride with propionic acid (8CI), Acetic propionic anhydride, Acetyl propionate |
| InChI formel | InChI=1S/C5H8O3/c1-3-5(7)8-4(2)6/h3H2,1-2H3 |
| SMIL | CCC(=O)OC(=O)C |
| IUPAC navn | acetyl propanoate |
| Molekylvægt (g/mol) | 116.1152 |
| Formel vægt | 116.0473 |