Få mere at vide
Acetyl-L-threonine, TRC
Artikelnummer. 30137772
Skift visning
Mængde:
50 mg
100 mg
500 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30137772 500 mg
30135643 100 mg
30009086 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Artikelnummer. 30137772 Leverandør Toronto Research Chemicals Leverandørnr. TRCA192013500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 17093-74-2
Molekylær formel C6H11NO4
Molekylvægt (g/mol) 161.16
Synonym L-Threonine, N-acetyl- (9CI, ACI), N-Acetyl-L-threonine (ACI), Threonine, N-acetyl-, L- (8CI), (2S,3R)-2-Acetamido-3-hydroxybutanoic acid, N-Acetylthreonine
IUPAC navn (2S,3R)-2-acetamido-3-hydroxybutanoic acid
SMIL C[C@H]([C@@H](C(=O)O)NC(=O)C)O
Tekniske data
Kemisk navn eller materiale Acetyl-L-threonine
InChI formel InChI=1S/C6H11NO4/c1-3(8)5(6(10)11)7-4(2)9/h3,5,8H,1-2H3,(H,7,9)(H,10,11)/t3-,5+/m1/s1
Mængde 500 mg
Formel vægt 161.0688