Få mere at vide
Acetyl-L-phenylalanine Ethyl Ester, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acetyl-L-phenylalanine Ethyl Ester |
| CAS | 2361-96-8 |
| Molekylær formel | C13H17NO3 |
| Mængde | 10 g |
| Synonym | N-Acetyl-L-phenylalanine Ethyl Ester, L-N-Acetyl-3-phenylalanine Ethyl Ester, APEE, Acetylphenylalanine Ethyl Ester, Ethyl N-Acetyl-L-phenylalaninate, L-N-Acetylphenylalanine Ethyl Ester, N-Acetylphenylalanine Ethyl Ester |
| InChI formel | InChI=1S/C13H17NO3/c1-3-17-13(16)12(14-10(2)15)9-11-7-5-4-6-8-11/h4-8,12H,3,9H2,1-2H3,(H,14,15)/t12-/m0/s1 |
| SMIL | CCOC(=O)[C@H](Cc1ccccc1)NC(=O)C |
| IUPAC navn | ethyl (2S)-2-acetamido-3-phenylpropanoate |
| Molekylvægt (g/mol) | 235.28 |
| Formel vægt | 235.1208 |