Få mere at vide
Acetyl L-Carnitine-d3 Hydrochloride, TRC
Artikelnummer. 30314637
Skift visning
Mængde:
2.5 mg
Pakningsstørrelse:
2.5mg
Artikelnummer. Mængde unitSize
30314637 2.5 mg 2.5mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30314637 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1719922.5MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetyl L-Carnitine-d3 chloride
CAS 1334532-17-0
Molekylær formel C9 2H3 H15 N O4 . Cl
Mængde 2.5 mg
Synonym [(2R)-2-Acetyloxy-4-hydroxy-4-oxobutyl]-dimethyl-(trideuteriomethyl)azanium chloride, Acetyl L-Carnitine-d3 Hydrochloride, Acetyl L-Carnitine-d3 Chloride, L-CARNITINE:HCL, O-ACETYL (N-METHYL-D3)
InChI formel InChI=1S/C9H17NO4.ClH/c1-7(11)14-8(5-9(12)13)6-10(2,3)4;/h8H,5-6H2,1-4H3;1H/t8-;/m1./s1/i2D3;
SMIL [Cl-].[2H]C([2H])([2H])[N+](C)(C)C[C@@H](CC(=O)O)OC(=O)C
IUPAC navn [(2R)-2-acetyloxy-3-carboxypropyl]-dimethyl-(trideuteriomethyl)azanium;chloride
Molekylvægt (g/mol) 242.72
Formel vægt 242.1113