Få mere at vide
Acetyl-d3 Chloride, TRC
Artikelnummer. 30312571
Skift visning
Mængde:
250 mg
Artikelnummer. Mængde
30312571 250 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312571 Leverandør Toronto Research Chemicals Leverandørnr. TRCA164497250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetyl-d3 Chloride
CAS 19259-90-6
Molekylær formel C2 2H3 Cl O
Mængde 250 mg
Synonym Acetyl-d3 chloride (6CI, 7CI, 8CI, 9CI), Acetyl chloride-d3, Trideuteroacetyl chloride, Acetyl Chloride-D3
InChI formel InChI=1S/C2H3ClO/c1-2(3)4/h1H3/i1D3
SMIL [2H]C([2H])([2H])C(=O)Cl
IUPAC navn 2,2,2-trideuterioacetyl chloride
Molekylvægt (g/mol) 81.52
Formel vægt 81.0061