Få mere at vide
Acetophos, TRC
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acetophos |
| CAS | 2425-25-4 |
| Molekylær formel | C8 H17 O5 P S |
| Mængde | 25 mg |
| Synonym | Acetic acid, [(diethoxyphosphinyl)thio]-, ethyl ester (9CI), Acetic acid, mercapto-, ethyl ester, O,O-diethyl phosphorothioate (6CI), Acetic acid, mercapto-, ethyl ester, S-ester with O,O-diethyl phosphorothioate (7CI,8CI), Acetaphos, Acetophos, Acetoxon, O,O-Diethyl S-(carbethoxy)methyl phosphorothiolate, O,O-Diethyl S-ethoxycarbonylmethyl phosphorothioate, Phosphorothioic acid, O,O-diethyl ester, S-ester with ethyl mercaptoacetate |
| InChI formel | InChI=1S/C8H17O5PS/c1-4-11-8(9)7-15-14(10,12-5-2)13-6-3/h4-7H2,1-3H3 |
| SMIL | CCOC(=O)CSP(=O)(OCC)OCC |
| IUPAC navn | ethyl 2-diethoxyphosphorylsulfanylacetate |
| Molekylvægt (g/mol) | 256.26 |
| Formel vægt | 256.0534 |