Få mere at vide
Acetophenone-d8, TRC
Artikelnummer. 30324914
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30324914 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30324914 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16401810MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetophenone-d8
CAS 19547-00-3
Molekylær formel C8 2H8 O
Mængde 10 mg
Synonym Ethanone-2,2,2-d3, 1-(phenyl-2,3,4,5,6-d5)- (ACI), 1-(Phenyl-2,3,4,5,6-d5)ethanone-2,2,2-d3 (ACI), Acetophenone-d8 (8CI), Ethanone-2,2,2-d3, 1-(phenyl-d5)- (9CI), 2,2,2-Trideuterio-1-(2,3,4,5,6-pentadeuteriophenyl)ethanone, Acetophenone-D8
InChI formel InChI=1S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6H,1H3/i1D3,2D,3D,4D,5D,6D
SMIL [2H]c1c([2H])c([2H])c(C(=O)C([2H])([2H])[2H])c([2H])c1[2H]
IUPAC navn 2,2,2-trideuterio-1-(2,3,4,5,6-pentadeuteriophenyl)ethanone
Molekylvægt (g/mol) 128.2
Formel vægt 128.1077