Få mere at vide
Acetophenone-d8, CDN
Artikelnummer. 30326648
Skift visning
Mængde:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30326648 5 g
30313100 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30326648 Leverandør CDN Leverandørnr. CDND29675G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetophenone-d8
CAS 19547-00-3
Molekylær formel C8 2H8 O
Mængde 5 g
InChI formel InChI=1 S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6 H,1H3/i1D3,2 D,3 D,4 D,5 D,6 D
SMIL [2 H]c1c([2 H])c([2 H])c(C(=O)C([2 H])([2 H])[2 H])c([2 H])c1[2 H]
IUPAC navn 2,2,2-trideuterio-1-(2,3,4,5,6-pentadeuteriophenyl)ethanone
Molekylvægt (g/mol) 128.2
Formel vægt 128.1077 g/mol
Procent renhed 98 atom % D, min 98% Chemical Purity