Få mere at vide
Acetophenone-d3, TRC
Artikelnummer. 30312765
Skift visning
Mængde:
1 g
Artikelnummer. Mængde
30312765 1 g
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312765 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1640231G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetophenone-d3
CAS 17537-31-4
Molekylær formel C8 D3 H5 O
Mængde 1 g
Synonym Ethanone-2,2,2-d3, 1-phenyl- (9CI, ACI), 1-Phenylethanone-2,2,2-d3 (ACI), Acetophenone-2,2,2-d3 (7CI, 8CI), 2,2,2-Trideuterio-1-phenylethanone, Aceto-d3-phenone, Acetophenone-α,α,α-d3, Acetophenone-d3, Acetophenone-2,2,2-D3, Acetophenone-d3 (methyl-d3)
InChI formel InChI=1S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6H,1H3/i1D3
SMIL [2H]C([2H])([2H])C(=O)c1ccccc1
IUPAC navn 2,2,2-trideuterio-1-phenylethanone
Molekylvægt (g/mol) 123.167
Formel vægt 123.076