Få mere at vide
Acetophenazine-d4 Dimaleate, TRC
Artikelnummer. 30312860
Skift visning
Mængde:
1 mg
Artikelnummer. Mængde
30312860 1 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30312860 Leverandør Toronto Research Chemicals Leverandørnr. TRCA1639321MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetophenazine-d4 Dimaleate
CAS 1331636-00-0
Molekylær formel C31H33D4N3O10S
Mængde 1 mg
Synonym 1-[10-[3-[4-(2-Hydroxyethyl-d4)-1-piperazinyl]propyl]-10H-phenothiazin-2-yl]ethanone (2Z)-2-Butenedioate, Acetophenazine-d4 Maleate, NSC 169180-d4, NSC 70600-d4, Phentoxate-d4, Sch 6673-d4, Tindal-d4, Tindala-d4
InChI formel InChI=1S/C23H29N3O2S.2C4H4O4/c1-18(28)19-7-8-23-21(17-19)26(20-5-2-3-6-22(20)29-23)10-4-9-24-11-13-25(14-12-24)15-16-27;2*5-3(6)1-2-4(7)8/h2-3,5-8,17,27H,4,9-16H2,1H3;2*1-2H,(H,5,6)(H,7,8)/b;2*2-1+/i15D2,16D2;;
SMIL O=C(O)/C=C\C(O)=O.O=C(C)C1=CC2=C(C=C1)SC3=CC=CC=C3N2CCCN4CCN(C([2H])([2H])C([2H])([2H])O)CC4
IUPAC navn (E)-but-2-enedioic acid;1-[10-[3-[4-(1,1,2,2-tetradeuterio-2-hydroxyethyl)piperazin-1-yl]propyl]phenothiazin-2-yl]ethanone
Molekylvægt (g/mol) 647.73