missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Acetonitrile, puriss. p.a., ACS Reagent, Reag. Ph. Eur., ≥99.5% (GC), Honeywell Riedel-de Haën™
Click to view available options
Quantity:
4 x 2.5 L
6 x 1 L
155 kg
Packaging:
Glass Bottle
Metal Drum
Description
ACS GENERAL USE SOLVENTS
- value priced solution for applications such as preparative-scale HPLC, cleaning and organic synthesis
- suitable for less demanding applications such as routine isocratic HPLC
- variations and decrease issues traceable to solvent contamination
- all solvents are tested to meet ACS general use specifications
Specifications
Melting Point | -48°C |
Acidity | Free acid (as CH3COOH): ≤0.001% |
Alkalinity | Free alkali (as NH3): ≤0.001% |
Linear Formula | CH3CN |
UN Number | UN1648 |
Beilstein | 741857 |
Formula Weight | 41.05g/mol |
Percent Purity | ≥99.5% (GC) |
Grade | ACS/puriss. p.a. |
Density | 0.786g/mL at 25°C |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'