Learn More
Acetonitrile, Hypergrade, For LC-MS LiChrosolv™, MilliporeSigma™
Product Code. 15833781
Change view
Click to view available options
Quantity:
1 L
2.5 L
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15833781 2.5 L 2.5 L
15823781 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15833781 Supplier Supelco Supplier No. 1000292500
Supelco Essentials Are in Stock
Use promo code 27958
View all promotions
 Restricted Item
På lager
Chemical Identifiers
CAS 75-05-08
Molecular Weight (g/mol) 41.05
Specifications
Melting Point -45.7°C
Acidity ≤ 0.0001 meq/g
Alkalinity ≤ 0.0002 meq/g
Density 0.78 g/cm3 at 20 °C
Quantity 2.5 L
Explosion Limit 3 to 17% (V)
Flash Point 2 °C
Ignition Point 524 °C
Packaging Glass Bottle
Residue after Evaporation ≤ 1 mg/L
Vapor Pressure 98.64 hPa at 20 °C
Water ≤ 0.01%
Synonym ACN, Methyl cyanide, Ethyl nitrile, Cyranomethane
Boiling Point 81.6°C
Percent Purity ≥99.97%
Grade Hypergrade
Chemical Name or Material Acetonitrile