Learn More
Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™ Product Code.: 15652550
Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™
BIOPHARMA
Product Code. 15652550
4 x 2.5 L, Glass Bottle, Pack of 4
Click to view available options
Quantity:
4 x 2.5 L
4 x 5 L
6 x 1 L
Packaging:
Glass Bottle
Poly Bottle
Unit Size:
Pack of 4
Pack of 6
Product Code. 15652550
Brand: Honeywell-Riedel-de Haen 242014X2.5LGL
Request Bulk or Custom Format
Puriss., meets analytical specification of BP, NF, Ph. Eur.,≥99% (GC)
  • Honeywell Riedel-de Haen has over 200 years of history in producing high-purity chemicals
  • It is the leading brand offering High-Purity Solvents for dedicated applications.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67-64-1
Molecular Formula C3H6O
Molecular Weight (g/mol) 58.08
MDL Number MFCD00008765
InChI Key CSCPPACGZOOCGX-UHFFFAOYSA-N
Synonym acetone, 2-propanone, propanone, dimethyl ketone, methyl ketone, dimethylformaldehyde, pyroacetic ether, beta-ketopropane, dimethylketal, chevron acetone
PubChem CID 180
ChEBI CHEBI:15347
IUPAC Name propan-2-one
SMILES CC(C)=O
Specifications
Melting Point -95°C
Color Colorless
Density 0.791g/mL at 25°C, 0.790 to 0.792g/mL at 20°C
Boiling Point 55°C to 57°C
Quantity 4 x 2.5 L
Explosion Limit 0.132
Flash Point −17°C (1.4°F)
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3COCH3
Vapor Pressure 184mmHg (20°C)
UN Number UN1090
Beilstein 635680
Formula Weight 58.08g/mol
Percent Purity ≥99% (GC)
Grade puriss.
Chemical Name or Material Acetone, Meets analytical specification of Ph. Eur.BP, NF
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot NumberN0530 Certificate TypeCertificate of Analysis Date05/04/2023 Product Code24201-2.5L,24201-2.5L-GL,24201-20KG,24201-25L,24201-4X2.5L,24201-4X2.5L-GL,24201-4X5L,24201-5L,24201-155KG,24201-6X1L,24201-1L
Lot NumberM0240 Certificate TypeCertificate of Analysis Date30/03/2023 Product Code24201-2.5L,24201-2.5L-GL,24201-20KG,24201-25L,24201-4X2.5L,24201-4X2.5L-GL,24201-4X5L,24201-5L,24201-155KG,24201-6X1L,24201-1L
Lot NumberM2840 Certificate TypeCertificate of Analysis Date30/03/2023 Product Code24201-2.5L,24201-2.5L-GL,24201-20KG,24201-25L,24201-4X2.5L,24201-4X2.5L-GL,24201-4X5L,24201-5L,24201-155KG,24201-6X1L,24201-1L
Lot NumberM2170 Certificate TypeCertificate of Analysis Date30/03/2023 Product Code24201-2.5L,24201-2.5L-GL,24201-20KG,24201-25L,24201-4X2.5L,24201-4X2.5L-GL,24201-4X5L,24201-5L,24201-155KG,24201-6X1L,24201-1L
Lot NumberM1670 Certificate TypeCertificate of Analysis Date30/03/2023 Product Code24201-2.5L,24201-2.5L-GL,24201-20KG,24201-25L,24201-4X2.5L,24201-4X2.5L-GL,24201-4X5L,24201-5L,24201-155KG,24201-6X1L,24201-1L
compliance-icons compliance-icons
Product Identifier
  • Acetone
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Product Content Correction
Product Title
Acetone, Puriss., meets analytical spec. of BP, NF, Ph. Eur., ≥99% (GC), Honeywell Riedel-de Haën™ >