Få mere at vide
Acetone 2,4-Dinitrophenylhydrazone-13C3, TRC
Artikelnummer. 30327673
Skift visning
Mængde:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30327673 50 mg
30322874 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30327673 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16385350MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetone 2,4-Dinitrophenylhydrazone-13C3
CAS 395075-27-1
Molekylær formel C613C3H10N4O4
Mængde 50 mg
Synonym 2-Propanone-13C3 2-(2,4-Dinitrophenyl)hydrazone, 2-Propanone-13C3 (2,4-Dinitrophenyl)hydrazone, Acetone-13C3 (2,4-Dinitrophenyl)hydrazone, 1-(2,4-Dinitrophenyl)-2-(propan-2-ylidene)hydrazine-13C3, NSC 131450-13C3, NSC 6120-13C3
InChI formel InChI=1S/C9H10N4O4/c1-6(2)10-11-8-4-3-7(12(14)15)5-9(8)13(16)17/h3-5,11H,1-2H3/i1+1,2+1,6+1
SMIL [13CH3]/[13C]([13CH3])=N/NC1=C([N+]([O-])=O)C=C([N+]([O-])=O)C=C1
IUPAC navn 2,4-dinitro-N-((1,2,3-13C3)propan-2-ylideneamino)aniline
Molekylvægt (g/mol) 241.18