Få mere at vide
Aceto-d3-phenone, CDN
Artikelnummer. 30319765
Skift visning
Mængde:
5 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30319765 10 g
30310140 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30319765 Leverandør CDN Leverandørnr. CDND009110G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetophenone-d3
CAS 17537-31-4
Molekylær formel C8 D3 H5 O
Mængde 10 g
Synonym Ethanone-2,2,2-d3, 1-phenyl- (9 CI, ACI), 1-Phenylethanone-2,2,2-d3 (ACI), Acetophenone-2,2,2-d3 (7 CI, 8 CI), 2,2,2-Trideuterio-1-phenylethanone, Aceto-d3-phenone, Acetophenone-α,α,α-d3, Acetophenone-d3, Acetophenone-2,2,2-D3, Acetophenone-d3 (methyl-d3)
InChI formel InChI=1 S/C8H8O/c1-7(9)8-5-3-2-4-6-8/h2-6 H,1H3/i1D3
SMIL [2 H]C([2 H])([2 H])C(=O)c1ccccc1
IUPAC navn 2,2,2-trideuterio-1-phenylethanone
Molekylvægt (g/mol) 123.167
Formel vægt 123.076 g/mol
Procent renhed 99 atom % D, min 98% Chemical Purity