Få mere at vide
Acetildenafil-d8, TRC
Artikelnummer. 30310027
Skift visning
Mængde:
10 mg
Artikelnummer. Mængde
30310027 10 mg
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30310027 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16160210MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetildenafil-d8
CAS 1189944-10-2
Molekylær formel C25H26D8N6O3
Mængde 10 mg
InChI formel InChI=1S/C24H32N6O3/c1-5-7-18-21-22(29(4)27-18)24(32)26-23(25-21)17-14-16(8-9-20(17)33-6-2)19(31)15-30-12-10-28(3)11-13-30/h8-9,14H,5-7,10-13,15H2,1-4H3,(H,25,26,32)/i10D2,11D2,12D2,13D2
SMIL [2H]C1([2H])N(CC)C([2H])([2H])C([2H])([2H])N(CC(=O)c2ccc(OCC)c(c2)C3=Nc4c(CCC)nn(C)c4C(=O)N3)C1([2H])[2H]
IUPAC navn 5-[2-ethoxy-5-[2-(2,2,3,3,5,5,6,6-octadeuterio-4-methylpiperazin-1-yl)acetyl]phenyl]-1-methyl-3-propyl-6H-pyrazolo[4,3-d]pyrimidin-7-one
Molekylvægt (g/mol) 474.63