missing translation for 'onlineSavingsMsg'
Learn More

Acetic hydrazide, 96%, Thermo Scientific Chemicals

Catalog Number p-7081139
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
1068-57-1
C2H6N2O
74.083
MFCD00007610
OFLXLNCGODUUOT-UHFFFAOYSA-N
acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
14039
CHEBI:48978
acetohydrazide
CC(=O)NN
This item is not returnable. View return policy
1068-57-1
C2H6N2O
74.083
MFCD00007610
OFLXLNCGODUUOT-UHFFFAOYSA-N
acethydrazide, acetylhydrazine, acetic hydrazide, acetic acid hydrazide, acetyl hydrazide, monoacetylhydrazine, n-acetylhydrazine, ethanehydrazonic acid, acetic acid, hydrazide, acetyl hydrazine
14039
CHEBI:48978
acetohydrazide
CC(=O)NN

Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Acetic hydrazide is used as an antibiotic for the treatment of mycobacterium tuberculosis. It is also involved in the preparation of sunitinib b reacting with 5-fluoroisatin. Further, it inhibits mycolic acid biosynthesis. It serves as an important intermediate in organic synthesis such as nitric furosemide Long (nifuratrone) in medicine.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with strong bases.
TRUSTED_SUSTAINABILITY
Melting Point ∼60°C to 68°C
Boiling Point 128°C to 129°C (18 mmHg)
Flash Point >110°C (230°F)
Quantity 25 g
Beilstein 506165
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 74.08
Percent Purity 96%
Chemical Name or Material Acetic hydrazide
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Germ cell mutagenicity Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H301-Toxic if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H341-Suspected of causing genetic defects.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'