Learn More
Acetic-d3 acid-d, for NMR, 99.5% atom D, Thermo Scientific Chemicals
Catalog Number p-3734286
290.64 DKK valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 mL
25 mL
50 mL
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1186-52-3
C2H4O2
64.08
MFCD00051051
QTBSBXVTEAMEQO-GUEYOVJQSA-N
acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
2723903
deuterio 2,2,2-trideuterioacetate
[2H]OC(=O)C([2H])([2H])[2H]
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
1186-52-3
C2H4O2
64.08
MFCD00051051
QTBSBXVTEAMEQO-GUEYOVJQSA-N
acetic acid-d4, acetic-d3 acid-d, 2h3 acetic 2h acid, tetradeuteroacetic acid, 2 h? ethan 2 h oic acid, acetic acid d4, 2h4 acetic acid, cd3cood, acetic acid d, acetic acid-d4, 99.5 atom % d
2723903
deuterio 2,2,2-trideuterioacetate
[2H]OC(=O)C([2H])([2H])[2H]
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For NMR
TRUSTED_SUSTAINABILITY
Physical Form Liquid
Color Colorless
Density 1.1300g/mL
Deuterium (D) 99.4% min.
Flash Point 40°C
Packaging Glass Bottle
Refractive Index 1.3709
Specific Gravity 1.13
Water 0.1% max. (Coulometric)
Quantity 5 mL
Melting Point 15.0°C to 16.0°C
Boiling Point 115.5°C
Linear Formula CD3CO2D
Beilstein 02, III, 203
Formula Weight 64.08
Grade NMR
Chemical Name or Material Acetic-d3 acid-d, 99.5 atom%D
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Acetic-d3 acid-d
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 A
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.