Få mere at vide
Acetic acid-nonyl ester, Dr. Ehrenstorfer
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Acetic Acid-Nonyl Ester |
| CAS | 143-13-5 |
| Analyt- eller komponentnavne | Acetic Acid-Nonyl Ester |
| Forsendelsestilstand | +20°C |
| Molekylær formel | C11 H22 O2 |
| Certificeringer/compliance | ISO/IEC 17025 |
| Synonym | Nonyl alcohol, acetate (6CI), 1-Acetoxynonane, 1-Nonyl acetate, Acetic acid n-nonyl ester, n-Nonanyl acetate, n-Nonyl acetate, n-Nonyl ethanoate, Nonanol acetate, Nonyl acetate, NSC 82356, Pelargonyl acetate |
| InChI formel | InChI=1S/C11H22O2/c1-3-4-5-6-7-8-9-10-13-11(2)12/h3-10H2,1-2H3 |
| SMIL | CCCCCCCCCOC(=O)C |
| IUPAC navn | nonyl acetate |
| Vis mere |