Få mere at vide
Acetic Acid, Glacial 25L, Solveco
Beskrivelse
Acetic acid 99%, also known as glacial acetic acid, in technical grade quality. This highly concentrated acid is commonly used in a wide range of industrial and chemical processes, including the manufacture of synthetic fibers, plastics, and coatings, as well as for pH adjustment, descaling, and as a reagent in laboratory settings.
Its high purity and reactivity make it a versatile component in both large-scale production and technical applications where concentration and consistency are critical.
Tekniske data
Tekniske data
| Kemisk navn eller materiale | Ethanoic acid |
| CAS | 64-19-7 |
| Molekylvægt (g/mol) | 60.05 g/mol |
| Procent renhed | 99.90% |
| Grad | Technical |
| Mængde | 25 L |
| Emballage | 15 L HDPE Container |
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.