Få mere at vide
Acetic Acid 3-Buten-1-yl Ester, TRC
Artikelnummer. 30318539
Skift visning
Mængde:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30318539 500 mg
30315609 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Artikelnummer. 30318539 Leverandør Toronto Research Chemicals Leverandørnr. TRCA158600500MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Tekniske data
Kemisk navn eller materiale Acetic Acid 3-Buten-1-yl Ester
CAS 1576-84-7
Molekylær formel C6H10O2
Mængde 500 mg
Synonym 1-Acetate-3-buten-1-ol, 1-Acetoxy-3-butene, 2-Vinylethyl Acetate, 3-Butenyl Acetate, 4-Acetoxy-1-butene, NSC 157648, 3-Buten-1-ol Acetate
InChI formel InChI=1S/C6H10O2/c1-3-4-5-8-6(2)7/h3H,1,4-5H2,2H3
SMIL CC(=O)OCCC=C
IUPAC navn but-3-enyl acetate
Molekylvægt (g/mol) 114.14
Formel vægt 114.0681