Få mere at vide
Acetic Acid-2-13C-2,2,2-d3, TRC
Artikelnummer. 30179308
Skift visning
Mængde:
25 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179308 250 mg
30179313 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179308 Leverandør Toronto Research Chemicals Leverandørnr. TRCA167644250MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 79562-15-5
Molekylær formel C13CHD3O2
Molekylvægt (g/mol) 64.06
Synonym Aci-Jel-2-13C,d3, E 260-2-13C,d3, Ethanoic Acid-2-13C,d3, Ethanoic Acid-2-13C,d3 Monomer, Ethylic Acid-2-13C,d3, Glacial Acetic-2-13C,d3 Acid, Methanecarboxylic Acid-2-13C,d3, NSC 111201-2-13C,d3, NSC 112209-2-13C,d3, NSC 115870-2-13C,d3, NSC 127175-2-13C,d3, NSC 132953-2-13C,d3, NSC 406306-2-13C,d3, Vinegar Acid-2-13C,d3;
IUPAC navn 2,2,2-trideuterioacetic acid
SMIL [2H][13C]([2H])([2H])C(=O)O
Tekniske data
Kemisk navn eller materiale Acetic Acid-2-13C-2,2,2-d3
Mængde 250 mg
InChI formel InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1+1D3
Formel vægt 64.0433