Få mere at vide
Acetic Acid-1-13C-2,2,2-d3, TRC
Artikelnummer. 30179459
Skift visning
Mængde:
50 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Mængde
30179459 50 mg
30179077 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Artikelnummer. 30179459 Leverandør Toronto Research Chemicals Leverandørnr. TRCA16764350MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Kemiske identifikatorer
CAS 63459-47-2
Molekylær formel 13C C H D3 O2
Molekylvægt (g/mol) 64.06
Synonym Aci-Jel-1-13C,d3, E 260-1-13C,d3, Ethanoic Acid-1-13C,d3, Ethanoic Acid-1-13C,d3 Monomer, Ethylic Acid-1-13C,d3, Glacial Acetic-1-13C,d3 Acid, Methanecarboxylic Acid-1-13C,d3, NSC 111201-1-13C,d3, NSC 112209-1-13C,d3, NSC 115870-1-13C,d3, NSC 127175-1-13C,d3, NSC 132953-1-13C,d3, NSC 406306-1-13C,d3, Vinegar Acid-1-13C,d3
IUPAC navn 2,2,2-trideuterioacetic acid
SMIL [2H]C([2H])([2H])[13C](=O)O
Tekniske data
Kemisk navn eller materiale Acetic acid-1-13C-2,2,2-d3
Mængde 50 mg
InChI formel InChI=1S/C2H4O2/c1-2(3)4/h1H3,(H,3,4)/i1D3,2+1
Formel vægt 64.0433